missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Aprepitant
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | Aprepitant |
| Quantité | 250 mg |
| Solubilité | practically insoluble |
| Poids de formule | 534.43 |
| Forme physique | Powder |
| Type de produit | Reagent |
| Pureté | ≥97.5% (HPLC) |
| Clé InChI | ATALOFNDEOCMKK-BYYRLHKVSA-N |
| SOURIRES | C[C@@H](O[C@@H]1OCCN(CC2=NC(=O)NN2)[C@@H]1C1=CC=C(F)C=C1)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
| Nom de l’IUPAC | 5-{[(2S,3R)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl}-2,3-dihydro-1H-1,2,4-triazol-3-one |
| Afficher plus de résultats |
RUO – Research Use Only