missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aprepitant

Catalog No. AC465310010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465310010 1 g
AC465312500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465310010 Supplier Thermo Scientific Chemicals Supplier No. 465310010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 170729-80-3 | C23H21F7N4O3, C23 H21 F7 N4 O3 | 534.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170729-80-3
Molecular Formula C23 H21 F7 N4 O3

Specifications

Quantity 1 g
Solubility practically insoluble
Formula Weight 534.43
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key ATALOFNDEOCMKK-BYYRLHKVSA-N
SMILES C[C@@H](O[C@@H]1OCCN(CC2=NC(=O)NN2)[C@@H]1C1=CC=C(F)C=C1)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
IUPAC Name 5-{[(2S,3R)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl}-2,3-dihydro-1H-1,2,4-triazol-3-one
Molecular Weight (g/mol) 534.44
Chemical Name or Material Aprepitant
Show More Show Less

RUO – Research Use Only