Learn More
Fisherbrand™ Drosophila Supplies Petri Dishes
Ideal for embryo collection and nematode research
$365.47 - $368.59
Specifications
Sterility Sterile
Packaging Easy-to-open bags
Quantity 20/Sleeve, 500 Case
Height (Metric) 15 mm
Diameter (Metric) 60 mm
View More Specs
Products 2
Catalog Number Mfr. No. Vent Price Quantity & Availability  
Catalog Number Mfr. No. Vent Price Quantity & Availability  
AS4050
View Documents Promotion Details
Fisherbrand™
3305
Yes
Case of 500 Each for $368.59
Only null left
AS4051
View Documents Promotion Details
Fisherbrand™
3308
No
Case of 500 Each for $365.47
Only null left
Description
Description
Ideal for embryo collection and nematode research
  • Use with the AS-225 Bottle Adapter Ring
  • Engineered for optimum flatness and uniform thickness
  • Packaged sterile in easy-to-open bags
  • 20 per sleeve, 500 per case
Specifications
Specifications
Sterile
20/Sleeve, 500 Case
60 mm
Easy-to-open bags
15 mm