Learn More
Fisherbrand™ Drosophila Supplies Petri Dishes
Catalog No. AS4050
Change view
Click to view available options
Vent:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vent
AS4050 Yes
AS4051 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AS4050 Supplier Fisherbrand™ Supplier No. 3305
Ideal for embryo collection and nematode research
  • Use with the AS-225 Bottle Adapter Ring
  • Engineered for optimum flatness and uniform thickness
  • Packaged sterile in easy-to-open bags
  • 20 per sleeve, 500 per case
Specifications
Disposable Single-use
Shape Round
Sterility Sterile
Type Petri Dish
Packaging Easy-to-open bags
Quantity 20/Sleeve, 500 Case
Vent Yes
Height (Metric) 15 mm
Diameter (Metric) 60 mm