missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Total RNA Control (Human)

Catalog No. 4307281
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4307281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4307281 Supplier Applied Biosystems™ Supplier No. 4307281
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provided at a convenient reverse-transcription-ready concentration for all RT-PCR reactions

TaqMan™ Control Total RNA is from Raji cells (Burkitt's lymphoma), and is conveniently packaged in a PCR-ready concentration for use as a control template in RT-PCR reactions. The Human total RNA is provided at a concentration of 50ng/μL.

Order Info

Shipping Condition: Dry Ice

Specifications

Format Frozen
Content And Storage 100 μl Total Human RNA (at 50 ng/μl). Store at -15 to -25°C.
Species Human
Shipping Condition Dry Ice
Concentration 50 ng/μL
Label or Dye Unlabeled
Quantity 100 μL
In which buffer is the Total RNA Control (Human) (Cat. No. 4307281) suspended?

The the Total RNA Control (Human) (Cat. No. 4307281) is suspended in 1 X TE buffer.

For Research Use Only. Not for use in diagnostic procedures.