missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Reverse Transcription Reagents DFS Item

TaqMan Reverse Transcription Reagents provide the necessary reagents for reverse transcriptase-PCR (RT-PCR). Random hexamers, oligo (dT)16, and reverse primers are included for cDNA synthesis flexibility.

Supplier:  Applied Biosystems™ 4304134

 View more versions of this product

Catalog No. 4304134

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

TaqMan Reverse Transcription Reagents provide the necessary reagents for reverse transcriptase-PCR (RT-PCR). Random hexamers, oligo (dT)16, and reverse primers are included for cDNA synthesis flexibility. These reagents are designed for reverse transcription and PCR amplification of a specific target RNA from either total RNA or mRNA. The reagents in this kit facilitate two-step RT-PCR, which is performed in two separate reactions and is useful when detecting multiple transcripts from a single cDNA reaction or when storing a portion of the cDNA.

Advantages of TaqMan Reverse Transcription Reagents

  • Contains MultiScribe Reverse Transcriptase, a recombinant Moloney murine leukemia virus (MMLV) reverse transcriptase
  • Includes random hexamers and oligo (dT)16 for flexibility
  • Individual components provide versatility in assay setup
  • Compatible with two-step RT-PCR reactions

Using TaqMan Reverse Transcription Reagents

  • TaqMan Reverse Transcription Reagents provide all the necessary components for reverse transcription of RNA to cDNA
  • For two-step RT-PCR, random hexamers, oligo (dT)16, and sequence-specific reverse primers can all be used
  • The choice of primers for reverse transcription is best made after experimentally evaluating all three priming systems
Specifications

Specifications

50 U/μL
Kit
42°C
MultiScribe
Dry Ice
First-Strand cDNA
Separate components
Up to 5 kb

Includes 10 units. Each unit contains:

• MultiScribe Reverse Transcriptase, 100 μL (50 U/μL)
• RNase Inhibitor, 100 μL (20 U/μL)
• dNTP Mixture, 1 mL (2.5 mM each dNTP)
• Oligo (dT)16, 100 μL (50 μM)
• Random Hexamers, 100 μL (50 μM)
• 10X RT Buffer, 1.5 mL
• MgCl2 Solution, 1.5 mL

Store at –25°C to –15°C.

Reverse Transcription
2000 Reactions
Yes
Real Time PCR (qPCR)
2000 Reactions at 10 μL
Reverse Transcription
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.