missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ PreAmp Master Mix Kit

Catalog No. 4384267
Encompass
Change view
Click to view available options
Quantity:
40 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4384267 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4384267 Supplier Applied Biosystems™ Supplier No. 4384267
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TaqMan™ PreAmp Master Mix Kit contains TaqMan™ PreAmp Master Mix and TaqMan™ Gene Expression Master Mix.

  • Amplify cDNA targets equally without introducing bias
  • Analyze mRNA from any precious sample such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE)
  • Stretch as little as 1ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression Assays
  • Analyze up to 100 gene expression targets with minimal hands-on time

Order Info

Guaranteed minimum shelf life is 60 days.

Specifications

Content And Storage Contains 2X TaqMan PreAmp Master Mix (1 mL), TaqMan™ Gene Expression Master Mix (5 mL), and TaqMan™ PreAmp Master Mix Kit Quick Reference Card. Store master mixes at 2-8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System
Passive Reference Dye No ROX
Product Line TaqMan
Product Type PreAmp Master Mix Kit
Quantity 40 reactions
Sufficient For 200 Reactions
Hot Start Built-In Hot Start
Sample Type cDNA
Show More Show Less
Can I use preamplification with the TaqMan SNP Genotyping Assays?

There is a preamplification protocol included with our TaqMan Sample-to-SNP Kit. This is designed to work with a lysate sample.

How much template can I start with when using the TaqMan PreAmp Master Mix Kit?

The TaqMan PreAmp Master Mix Kit (P/N 4384267) is intended for use with very small quantities of cDNA (1 to 250 ng). This kit allows you to increase the quantity of specific cDNA targets for gene expression analysis using TaqMan Gene Expression Assays. Starting material is increased prior to PCR and the resulting preamplification product is then used for PCR. The TaqMan PreAmp Master Mix Kit enables multiplex preamplification of up to 100 targets at a time and provides unbiased amplification of specific amplicons for analysis with TaqMan Gene Expression Assays. For more detailed information on this kit, please refer to the TaqMan PreAmp Master Mix Kit Protocol (P/N 4384557).

What is the TaqMan PreAmp Master Mix Kit?

The TaqMan PreAmp Master Mix Kit (P/N 4384267) preamplifies small amounts of cDNA without introducing amplification bias to the sample. Preamplification enables you to stretch your limited sample into many more real-time PCR reactions. TaqMan PreAmp Master Mix Kit consists of TaqMan PreAmp Master Mix and the TaqMan Gene Expression Master Mix. The two work hand in hand to provide the best results in preamplification and quantitation of mRNA.

For more detailed information on this kit, please refer to the TaqMan PreAmp Master Mix Kit Protocol (P/N 4384557).

How does the SuperScript IV Single Cell/Low Input cDNA PreAmp Kit differ from TaqMan PreAmp Master Mix?

TaqMan PreAmp Master Mix is designed for targeted preamplification of up to 100 targets from small amounts of cDNA, while SuperScript IV Single Cell/Low Input cDNA PreAmp Kit allows global preamplification of full-length cDNA from single cells or low input RNA. It provides reagents for cell lysis, reverse transcription and preamplification steps.

For Research Use Only. Not for use in diagnostic procedures.