missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Control Genomic DNA (human)

Catalog No. 4312660
Encompass
Change view
Click to view available options
Quantity:
2 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4312660 2 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4312660 Supplier Applied Biosystems™ Supplier No. 4312660
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conveniently packaged in a PCR-ready concentration for use as a control template in PCR reactions

The Human gDNA (male) is provided in two tubes (100μL each) at a concentration of 10ng/μL. TaqMan™ Control Genomic Human DNA is also included in several Applied Biosystems™ real-time PCR kits.

Order Info

Shipping Condition: Dry Ice

Recommended Storage

Store at -15° to -25°C.

Specifications

Format Frozen
Content And Storage Human gDNA (Male) packaged in two tubes (100μL each). Store at -15 to 25°C.
Species Human
Shipping Condition Dry Ice
Concentration 10 ng/μL
Label or Dye Unlabeled
Quantity 2 Tubes
Product Line TaqMan

For Research Use Only. Not for use in diagnostic procedures.