missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SYBR™ Green Universal Master Mix

Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.

Supplier:  Applied Biosystems™ 4312704

Encompass

 View more versions of this product

Catalog No. 4312704


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Includes: 2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.

Description

Description

Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.

  • SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
  • AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
  • dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
  • Proprietary buffer enhancements ensure performance and reliability
  • Premixed components stored at 2° to 8°C significantly reduce assay setup time

Maximum Flexibility and Convenience:

  • Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
  • SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
  • The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
  • Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
  • This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
  • SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
  • Store at -20°C

Order Info

Shipping Condition: Wet Ice

Specifications

Chemical Identifiers

2X
Tube
qPCR
Standard
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
SYBR
High
AmpliTaq Gold DNA Polymerase
SYBR Green
SYBR
Gene Expression

Specifications

2X
SYBR
High
AmpliTaq Gold DNA Polymerase
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
2000 Reactions of 50 μL
2000 Reactions
10 x 5 mL
The 10 x 5 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 2000 reactions based on a 50 μl reaction volume.

Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Tube
qPCR
Standard
SYBR Green
SYBR
10 x 5 mL
Gene Expression
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.