missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical Adhesive Film Kit

Catalog No. 4313663
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4313663 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4313663 Supplier Applied Biosystems™ Supplier No. 4313663

Includes

MicroAmp Optical Adhesive Covers (quantity 20), Applicator (quantity 1), MicroAmp Optical Cover Compression Pad (quantity 1)

Designed to fit on MicroAmp Optical 96-well Reaction Plates

 

  • MicroAmp Optical Adhesive Film reduces the chance of well-to-well contamination and sample evaporation when applied to a microplate
  • MicroAmp Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films
  • MicroAmp Adhesive Film Applicator is used to form a tight seal between a microplate and an adhesive film
  • Pressure-sensitive adhesive provides a tight seal over every well
  • Will not interfere with sample reading
  • Press-N-Seal: Align properly to the plate, then press to seal
  • Easy to apply—does not stick to your glove

 

Prevent Loss of Precious Samples

  • Use this optically clear adhesive film to seal the samples into the wells of a microplate to reduce the possibility of cross-contamination between sample wells and help ensure consistent real-time PCR data
  • Easy to Use
  • Simply peel the backing away from the adhesive film and apply film firmly across the top of your microplate
  • Stack microplates and read samples as you would normally except with less chance of sample contamination
  • MicroAmp Optical Film Compression Pads are designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films
  • Use the MicroAmp Adhesive Film Applicator to form a tight seal between a microplate and the adhesive film
Form a Tight Seal
  • The MicroAmp Adhesive Film Applicator enables use to ergonomically push an adhesive film down on a microplate to form a tight seal and minimize air bubbles. This tight seal will prevent well-to-well contamination and sample loss
  • Quick and Easy
  • Use the MicroAmp Adhesive Film Applicator to speed the process of sealing multiple microplates
  • This tool is designed according to the microplate dimensions so microplate can be quickly sealed
  • The rugged proprietary plastic endures constant use for large screening applications

 

See user's manual or package insert for limited label license and trademark information.

PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room Temperature

Specifications

Color Optical
Material Polyester
No. of Wells 96
Quantity 1 kit
Content And Storage Includes: MicroAmp™ Optical Adhesive Covers (quantity 20), Applicator (quantity 1), MicroAmp™ Optical Cover Compression Pad (quantity 1)
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7900HT System, GeneAmp 9700, QuantStudio™, ViiA™ 7 Dx System, ViiA™ 7 System, QuantStudio™ Dx
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
Product Line MicroAmp
Adhesive Pressure Sensitive
Plate Compatibility 96-well Plate
Show More Show Less
How do I correctly apply MicroAmp Optical Adhesive Covers to an optical 96- or 384-well plate?

Please follow the instructions below for the proper application of MicroAmp Optical Adhesive Covers (Cat. Nos. 4311971, 4313663 and 4360954).

The MicroAmp Optical Adhesive Cover uses a pressure-sensitive adhesive backing to adhere the cover to the optical 96- or 384-well plate. It is imperative that you use enough force to activate the pressure-sensitive adhesive to prevent evaporation from the wells.
Please note: Users should not expect the Optical Adhesive Cover to be very sticky to the touch on the side that adheres to the optical plate.

The correct way to apply the cover is as follows:

1. Apply firm pressure when using the Adhesive Seal Applicator (Cat. No. 4333183) to press the cover onto the plate.

2. Run the applicator firmly from left to right several times. Run the applicator up and down several times.

3. Run the edge of the applicator around the border (following the red line outside the wells) of the cover.

4. Repeat the left to right and up and down motions to assure a good final seal.

5. Inspect each well to make sure that each well edges are imprinted on the optical adhesive cover.
Please Note: Using the Applicator's edge to seal between the wells is NOT recommended. It may actually pull the seal off the wells.

Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.