missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 48-Well Optical Adhesive Film

Catalog No. 4375928
Encompass
Change view
Click to view available options
Quantity:
100 films
25 films
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4375928 25 films
4375323 100 films
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4375928 Supplier Applied Biosystems™ Supplier No. 4375928

Includes

100 or 25 MicroAmp 48-Well Optical Adhesive Films

Reduces the chance of well-to-well contamination and sample evaporation when applied to a 48-well microplate

  • Pressure-sensitive adhesive provides tight seal over every well
  • Will not interfere with sample reading
  • Easy to apply and remove, will not stick to gloves
  • Prevent Sample Loss
  • Optically clear adhesive film prevents sample loss by securely sealing into wells of microplates and reducing leaks, evaporation, and contamination between sample wells to help ensure consistent real-time PCR data
  • Easy to Use: simply peel backing and apply firmly across top of microplate
  • Once sealed, microplates can be safely stacked without worrying about contamination
  • Designed to fit MicroAmp Fast Optical 48-Well Plates
Note: See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.

Applications:

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Color Optical
Material Polyester
No. of Wells 48
Quantity 25 films
Content And Storage Contains 25 MicroAmp™ 48-Well Optical Adhesive Films. Store at room temperature.
For Use With (Equipment) StepOne™,
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
Product Line MicroAmp
Adhesive Pressure Sensitive
Plate Compatibility 48-well Plate
Show More Show Less
Can I cut an Optical Adhesive seal in half to fit the 48well StepOne system plate?

It is not recommended that you do so. Please use the MicroAmp 48-Well Optical Adhesive Film (P/N 4375928).

Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.