missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea)

Catalog No. N8080260
Encompass
Change view
Click to view available options
Quantity:
1 mL
6 x 1 mL
12 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8080260 1 mL
4303441 6 x 1 mL
4303442 12 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N8080260 Supplier Applied Biosystems™ Supplier No. N8080260

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.

Key advantages of GeneAmp dNTPs

  • High purity dNTPs are ideal for use in PCR reactions or other applications
  • Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
  • Premixed dNTP blends are available in various concentrations, offering ease of use

Specifications

Concentration 10 mM
Content And Storage GeneAmp dNTP Blend, 2.5 mM (1 mL)

Store in a constant temperature freezer at –20°C.
Form Liquid
Product Line GeneAmp
Quantity 1 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Why do some dNTP mixes contain dTTP and others TTP?

Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions

For Research Use Only. Not for use in diagnostic procedures.