missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APOF Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5118960

 Afficher plus de versions de ce produit

Numéro de catalogue. PIPA5118960

Il en reste null

Description

Description

APOF Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol.
Spécifications

Identifiants chimiques

APOF
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Mouse
APOF
AI255964; APOF; Apo-F; Apolipoprotein F; APON; Ba1-666; DKFZp781G18150; Leukemia virus-inactivating factor; lipid transfer inhibitor protein; Liver regeneration-related protein LRRG151; LTIP; LVIF; MGC22520; ovarian and testicular apolipoprotein N; Unknown (protein for MGC:137251)
APOF
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q91V80
103161
Recombinant protein Apolipoprotein F. The antigen corresponds to amino acid range 111-314 of the target protein.
Antibody

Spécifications

APOF
Polyclonal
Unconjugated
APOF
AI255964; APOF; Apo-F; Apolipoprotein F; APON; Ba1-666; DKFZp781G18150; Leukemia virus-inactivating factor; lipid transfer inhibitor protein; Liver regeneration-related protein LRRG151; LTIP; LVIF; MGC22520; ovarian and testicular apolipoprotein N; Unknown (protein for MGC:137251)
Rabbit
Antigen affinity chromatography, Protein A
RUO
103161
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q91V80
APOF
Recombinant protein Apolipoprotein F. The antigen corresponds to amino acid range 111-314 of the target protein.
200 μL
Primary
Mouse
Antibody
IgG
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions