missing translation for 'onlineSavingsMsg'
Learn More

Apigenin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002023
Change view
Click to view available options
Quantity:
75 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002023 75 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002023 Supplier Merck Emd Millipore Supplier No. PHR219875MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 520-36-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C15H10O5
MDL Number MFCD00006831
Melting Point >300°C
Synonym Apigenin; 4′,5,7-Trihydroxyflavone; 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone
Type Certified Reference Material
Quantity 75 mg
Formula Weight 270.24
Grade Certified Reference Material