missing translation for 'onlineSavingsMsg'
Learn More

Apigenin, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014431
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014431 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014431 Supplier Merck Emd Millipore Supplier No. 4225110MG
Only null left

Flavonoids belong to polyphenolic class of compounds. They play crucial role in biological activities which include tumor growth inhibition and chemoprevention. Apigenin is a plant flavonoid mostly used to inhibit ornithine decarboxylase activity and skin tumor.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 520-36-5
Physical Form Neat
Linear Formula C15H10O5
MDL Number MFCD00006831
Melting Point >300°C
Synonym 4 ′,5,7-Trihydroxyflavone; 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone
Type Analytical Standard
Quantity 10 mg
Formula Weight 270.24
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less