missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichlorure d’antimoine, réactif ACS
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Point d’ébullition | 223,0 °C |
| Nom ou substance chimique | Antimony trichloride |
| Point de fusion | 73,0 °C |
| Couleur | Blanc |
| Forme physique | Poudre, cristaux et/ou morceaux |
| Quantité | 500 g |
| Emballage | Bouteille en verre |
| Formule linéaire | SbCl3 |
| Fieser | 08,19 |
| Indice de Merck | 15,696 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Toxic to aquatic life with long lasting effects. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 233-047-2 |
| RTECSNumber | CC4900000 |
| TSCA | TSCA |
RUO – Usage réservé à la recherche