missing translation for 'onlineSavingsMsg'
Learn More

Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)

Catalog No. AA10095A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10095A3 2 kg
AA1009522 100 g
AA1009536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10095A3 Supplier Thermo Scientific Chemicals Supplier No. 010095A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-36-0 | Sb | 121.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]

Specifications

Color Silver to Gray
Appearance Solid in various forms
Quantity 2 kg
Odor Odorless
Assay Percent Range 99.5% (Metals basis)
IUPAC Name antimony
Chemical Name or Material Antimony
EINECSNumber 231-146-5
RTECSNumber CC4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only