Learn More
ACROBiosystems Inc Anti-SARS-CoV-2 Spike S1 Antibody, Chimeric mAb
Numéro de catalogue. 502019446
Encompass
Antibody;100UG;Human IgG1/kappa;The cross-reactivity with other coronaviruses has not been tested yet.;Anti-SARS-CoV-2 Spike S1 Antibody, Chimeric MAb (S1N-M13A1) is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified recombinant SARS-CoV-2 Spike S1 Protein.
It is recommended to use antibody (S1N-M13A1) and (S1N-M12A1) as a pair set to detect SARS-CoV-2 Spike S1 in ELISA or serological assays.; The protein has a calculated MW of 49.1 kDa & 23.5 kDa. ;Immobilized SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H2) at 2 g/mL (100 L/well) can bind Anti-SARS-CoV-2 Spike S1 Antibody, chimeric mAb (Cat. No. S1N-M13A1) with a linear range of 0.08-2.5 ng/mL (QC tested).
Numéro de catalogue. 502019446 Fournisseur ACROBiosystems Inc Code fournisseur S1NM13A1100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Antibody;100UG;Human IgG1/kappa;The cross-reactivity with other coronaviruses has not been tested yet.;Anti-SARS-CoV-2 Spike S1 Antibody, Chimeric MAb (S1N-M13A1) is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified recombinant SARS-CoV-2 Spike S1 Protein.
It is recommended to use antibody (S1N-M13A1) and (S1N-M12A1) as a pair set to detect SARS-CoV-2 Spike S1 in ELISA or serological assays.; The protein has a calculated MW of 49.1 kDa & 23.5 kDa. ;Immobilized SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H2) at 2 g/mL (100 L/well) can bind Anti-SARS-CoV-2 Spike S1 Antibody, chimeric mAb (Cat. No. S1N-M13A1) with a linear range of 0.08-2.5 ng/mL (QC tested).