missing translation for 'onlineSavingsMsg'
Learn More

TSC22 domain family, member 2, Mouse, Clone: 6G10, Abnova™

Numéro de catalogue. 89001512
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
89001512 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 89001512 Fournisseur Abnova Corporation Code fournisseur H00009819M01A
Il en reste null

Mouse monoclonal antibody raised against a partial recombinant TSC22D2.

Sequence: KMPAKKKSCFQITSVTTAQVATSITEDTESLDDPDESRTEDVSSEIFDVSRATDYGPEEVCERSSSEETLNNVGDAETPGTVS

Spécifications

Antigen TSC22 domain family, member 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6G10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TSC22D2.
Formulation ascites with no preservative
Gene TSC22D2
Gene Accession No. NM_014779
Gene Alias KIAA0669/TILZ4a/TILZ4b/TILZ4c
Gene Symbols TSC22D2
Host Species Mouse
Immunogen TSC22D2 (NP_055594, 3 a.a. ∼ 85 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9819
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Afficher plus de résultats Afficher moins de résultats