Learn More
ACROBiosystems Inc ACROBiosystems Anti-Trastuzumab Antibodies (recommended for PK/PD)
Numéro de catalogue. 502106934
Encompass
Antibody;1MG;IgG1 / kappa;Recognizes Trastuzumab specifically, no cross reactivity with other humanized antibodies.;Anti-Trastuzumab Antibodies (recommended for PK/PD) (TRB-Y1b) are expressed from human 293 cells (HEK293).;This protein . ;Immobilized Trastuzumab at 1 g/mL (100 L/well) can bind Anti-Trastuzumab Antibodies (Cat. No. TRB-Y1b) with a linear range of 0.4-6 ng/mL (QC tested);Immobilized Anti-Trastuzumab Antibodies (Cat. No. TRB-Y1b) at 2g/mL, add increasing concentrations of Trastuzumab in the 10% human serum and then add Biotinylated Anti-Trastuzumab Antibodies (recommended for PK/PD) (Cat. No. TRB-BY5) at 2g/mL. Detection was performed using HRP-conjugated streptavidin with sensitivity of 48 ng/mL (QC tested).
Numéro de catalogue. 502106934 Fournisseur ACROBiosystems Inc Code fournisseur TRBY1B1MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Antibody;1MG;IgG1 / kappa;Recognizes Trastuzumab specifically, no cross reactivity with other humanized antibodies.;Anti-Trastuzumab Antibodies (recommended for PK/PD) (TRB-Y1b) are expressed from human 293 cells (HEK293).;This protein . ;Immobilized Trastuzumab at 1 g/mL (100 L/well) can bind Anti-Trastuzumab Antibodies (Cat. No. TRB-Y1b) with a linear range of 0.4-6 ng/mL (QC tested);Immobilized Anti-Trastuzumab Antibodies (Cat. No. TRB-Y1b) at 2g/mL, add increasing concentrations of Trastuzumab in the 10% human serum and then add Biotinylated Anti-Trastuzumab Antibodies (recommended for PK/PD) (Cat. No. TRB-BY5) at 2g/mL. Detection was performed using HRP-conjugated streptavidin with sensitivity of 48 ng/mL (QC tested).