missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nuclear receptor 2C2-associated protein, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TRA16.
Supplier: Abnova Corporation H00126382A01
Description
Sequence: NSDQGPSQWVTLEFPQLIRVSQLQIQFQGGFSSRRGCLEGSQGTQALHKIVDFYPEDNNSLQTFPIPAAEVDRLKVTFEDATDFFGRVVIYHLRVLGEKVSpécifications
| nuclear receptor 2C2-associated protein | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TRA16. | |
| NR2C2AP | |
| TRA16 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 126382 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_176880 | |
| NR2C2AP | |
| TRA16 (NP_795361, 40 a.a. ∼ 139 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |