missing translation for 'onlineSavingsMsg'
Learn More

SV40 Large T Antigen Mouse anti-Sv40 Infected Cells, Clone: PAb416, MilliporeSigma™

Numéro de catalogue. MABF121MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MABF121MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MABF121MI Fournisseur MilliporeSigma Code fournisseur MABF121
Il en reste null

Mouse Monoclonal Antibody

Anti-SV40 Large T Antigen Antibody, clone PAb416 is an antibody against SV40 Large T Antigen for use in Western Blotting, immunocytochemistry.

Spécifications

Antigen SV40 Large T Antigen
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone PAb416
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NC_001669
Host Species Mouse
Immunogen Purified large T-antigen of SV40
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats

For Research Use Only