missing translation for 'onlineSavingsMsg'
Learn More

S100 calcium binding protein A5, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant S100A5.

Supplier:  Abnova Corporation H00006276A01

Numéro de catalogue. 89004302


Il en reste null

Description

Description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has a Ca2+ affinity 20- to 100-fold higher than the other S100 proteins studied under identical conditions. This protein also binds Zn2+ and Cu2+, and Cu2+ strongly which impairs the binding of Ca2+. This protein is expressed in very restricted regions of the adult brain. [provided by RefSeq]

Sequence: MPAAWILWAHSHSELHTVMETPLEKALTTMVTTFHKYSGREGSKLTLSRKELKELIKKELCLGEMKESSIDDLMKSLDKNSDQEIDFKEY
Spécifications

Spécifications

S100 calcium binding protein A5
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant S100A5.
S100A5
S100D
Mouse
50 μL
Yes
6276
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_002962
S100A5
S100A5 (NP_002953, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions