missing translation for 'onlineSavingsMsg'
Learn More

Phospho-tyrosine Mouse anti-All, HRP, Clone: 4G10™, MilliporeSigma™

Numéro de catalogue. 16105MI
Modifier l'affichage
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
16105MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 16105MI Fournisseur MilliporeSigma Code fournisseur 16 105
Il en reste null

Mouse Monoclonal Antibody

Anti-Phosphotyrosine Antibody, clone 4G10™, HRP conjugate detects level of Phosphotyrosine and has been published and validated for use in ELISA and western blotting.

Spécifications

Antigen Phosphotyrosine
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G10™
Conjugate HRP
Formulation 200μg of IgG2bκ-HRP conjugate in 100μL PBS containing 0.05% ProClin™150. Sufficient for 10 western blots, if used at the optimum concentration of 2μg/mL in 10mL blocking buffer. Liquid.
Host Species Mouse
Immunogen Phosphotyramine-KLH.
Purification Method Protein G-Sepharose™ chromatography
Quantity 1 Unit
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for 6 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Afficher plus de résultats Afficher moins de résultats

For Research Use Only