missing translation for 'onlineSavingsMsg'
Learn More

Phospho-serine Mouse anti-Vertebrates, Clone: 4A4, MilliporeSigma™

Numéro de catalogue. 051000MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
051000MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 051000MI Fournisseur MilliporeSigma Code fournisseur 051000
Il en reste null

Mouse Monoclonal Antibody

Detect Phosphoserine using this Anti-Phosphoserine Antibody, clone 4A4 (mouse IgG1) validated for use in ELISA, flow cytometry, immunofluoresence, immunohistochemistry (paraffin) and western blotting.

Spécifications

Antigen Phosphoserine
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4A4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing PBS with 0.1% sodium azide and 30% glycerol.
Host Species Mouse
Immunogen Phosphoserine coupled to KLH.
Purification Method Protein G-Sepharose™ chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats

For Research Use Only