missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKA-RII (Ser96) Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT58
Description
This Anti-phospho PKA-RII (Ser96) Antibody is validated for use in western blotting for the detection of phospho PKA-RII (Ser96).Spécifications
| phospho PKA-RII (Ser96) | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine, pH 7.4, 150mM NaCl, with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot | |
| Unconjugated | |
| PRKAR2A; PKR2; PRKAR2 | |
| KLH-conjugated linear peptide corresponding to mouse PKA-RIIalpha phosphorylated at Ser96. | |
| 100 μL | |
| NP_032950 | |
| 2-8°C for 12 months |
For Research Use Only