missing translation for 'onlineSavingsMsg'
Learn More

PATE1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Numéro de catalogue. ABC930MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ABC930MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. ABC930MI Fournisseur MilliporeSigma Code fournisseur ABC930
Il en reste null

Rabbit Polyclonal Antibody

This Anti-PATE1 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of PATE1.

Spécifications

Antigen PATE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PATE1; PATE
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human PATE1.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_612151
Target Species Human
Content And Storage 2-8°C for 12 months
Afficher plus de résultats Afficher moins de résultats

For Research Use Only