Learn More
Nuclei Mouse anti-Human Only, Clone: 235-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MAB1281
Numéro de catalogue. MAB1281MI
Il en reste null
Description
Description
Detect Nuclei using this Anti-Nuclei Antibody, clone 235-1 validated for use in immunohistochemistry (paraffin) and immunoprecipitation.
Spécifications
Spécifications
Nuclei
Monoclonal
Unconjugated
Mouse
100 μL
Human
Purified
Immunohistochemistry (Paraffin), Immunoprecipitation
235-1
Hybridoma supernatant. Liquid in PBS. Contains no preservative.
Protein A purfied
Primary
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
IgG1
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS