missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFκB p52 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06413
Description
Detect NFκB p52 with Anti-NFκB p52 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.Spécifications
| NFκB p52 | |
| Polyclonal | |
| Protein A Purified immunoglobulin in in 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% NaN3 as a preservative. | |
| Rabbit | |
| Unpurified | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| NFKB2; LYT10; LYT-10; H2TF1; Lyt10 | |
| KLH conjugated synthetic peptide (CYNPGLDGIIEYDDFK) contained within amino acids 4-19 of human NF κB, p52 | |
| 100 μL | |
| NM_019408 | |
| −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG |
For Research Use Only