missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuroD 6 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Description
This Anti-NeuroD 6 Antibody is validated for use in western blot, immunohistochemistry for the detection of NeuroD 6.
Spécifications
Spécifications
| Antigen | NeuroD 6 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. |
| Gene Symbols | NEUROD6; MATH2; Math-2; ATOH2; NeuroD6; NEX1M; Atoh2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Afficher plus de résultats |
For Research Use Only