missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MuSK Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Description
Anti-MuSK antibody is an antibody against MuSK for use in western blotting, immunohistochemistry and immunoprecipitation.
Spécifications
Spécifications
| Antigen | MuSK |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal serum with 0.05% sodium azide. |
| Gene Symbols | MUSK |
| Host Species | Rabbit |
| Immunogen | Recombinant protein corresponding to the extracellular domain of mouse MuSK. |
| Purification Method | Unpurified |
| Quantity | 100 μL |
| Afficher plus de résultats |
For Research Use Only