missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anti-miR™ hsa-let-7c miRNA Inhibitor Positive Control

Catalog No. 4392431
Encompass
Change view
Click to view available options
Quantity:
5 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4392431 5 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4392431 Supplier Invitrogen™ Supplier No. 4392431

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Anti-miR™ hsa-let-7c miRNA Inhibitor Positive Control

Invitrogen™ Anti-miR™ miRNA Inhibitors are chemically modified, single-stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA) molecules. These ready-to-use inhibitors can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. Anti-miR™ let-7c miRNA Inhibitor Positive Control provides a convenient, validated positive control for experiments using Anti-miR miRNA Inhibitors. Endogenous let-7 miRNA negatively regulates HMGA2, a ubiquitously expressed, nonhistone chromatin protein that modulates gene expression through changes in chromatin architecture. When transfected into human and mouse cells, Anti-miR hsa-let-7c miRNA Inhibitor blocks endogenous let-7c miRNA, resulting in increased levels of HMGA2 mRNA.

Accessory Products:
Also available are Anti-miR™ miRNA Inhibitors (SKU #AM17000 and #AM17001). To select your miRNA sequence and order, please visit the GeneAssist™ miRNA Workflow Builder. Anti-miR™ miRNA Inhibitor Negative Control #1 (SKU #AM17010) is a random sequence that has been validated to produce no identifiable effects on known miRNA function. Cy™3 and FAM™ dye-labeled Anti-miR Negative Controls (SKU #AM17011 and #AM17012, respectively) are also available. The Pre-miR™ miRNA Starter Kit (SKU #AM1540) can be used to optimize transfection of Pre-miR™ miRNA Precursors.

Specifications

Content And Storage Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
Control Type Positive Control
RNAi Type miRNA Inhibitor
For Use With (Application) miRNA & Non-Coding RNA Analysis
Label or Dye Unconjugated
Product Line Ambion, Anti-miR
Product Type miRNA Inhibitor
Quantity 5 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.