missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lck Rabbit anti-Bovine, Human, Mouse, Polyclonal, MilliporeSigma™
Description
Anti-Lck Antibody is an antibody against Lck for use in immunoprecipitation and western blotting.
Spécifications
Spécifications
| Antigen | Lck |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide |
| Gene Symbols | LCK; p56lck; YT16; pp58lck; p56-LCK; LSK |
| Host Species | Rabbit |
| Immunogen | GST fusion protein corresponding to amino acids 1-58 of human Lck |
| Purification Method | Protein A chromatography |
| Quantity | 200 μg |
| Afficher plus de résultats |
For Research Use Only