missing translation for 'onlineSavingsMsg'
Learn More

anti-IL6, HRH1 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89022988
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89022988 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89022988 Supplier Abnova Corporation Supplier No. DI0229

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

IL6 rabbit purified polyclonal antibody (20μg); HRH1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the IL6 protein, and the other against the HRH1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen IL6
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene IL6
Gene Alias BSF2/HGF/HSF/IFNB2/IL-6
Gene Symbols IL6
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3569/3269
Includes Antibody pair set content: 1. IL6 rabbit purified polyclonal antibody (20 ug) 2. HRH1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type IL6 & HRH1 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less