missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKKα Mouse anti-Human, Mouse, Monkey, Clone: 14A231, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05536
Description
Anti-IKKα Antibody, clone 14A231 is an antibody against IKKα for use in western blotting.Spécifications
| IKKα | |
| Monoclonal | |
| Unconjugated | |
| NM_001278 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 14A231 | |
| Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. | |
| CHUK, NFKBIKA, IKKA, IKBKA, IkBKA, TCF16, IKK1, IKK-A, IKK-alpha | |
| His-tagged, full length human IKKα | |
| 100 μg | |
| Human, Monkey, Mouse | |
| Purified |
For Research Use Only