Learn More
GSK3α/Β Rabbit anti-Human, Mouse, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier:  MilliporeSigma 05903
Numéro de catalogue. 05903MI
Il en reste null
Description
Description
Anti-GSK3α/β Antibody is an antibody against GSK3α/β for use in immunoprecipitation and western blotting.
Spécifications
Spécifications
GSK3α/β
Monoclonal
NM_019884/NM_002093
N-terminal 6His-tagged recombinant full length human GSK3β
100 μL
GSK3A, DKFZp686D0638
−20°C for 24 months
IgG
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Protein A purfied
Primary
Human, Mouse
Purified
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS