missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eukaryotic translation initiation factor 3, subunit H, Mouse, Clone: 3B12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EIF3S3.
Supplier: Abnova Corporation H00008667M01
Description
Sequence: YDPIKTAQGSLSLKAYRLTPKLMEVCKEKDFSPEALKKANITFEYMFEEVPIVIKNSHLINVLMWELEKKSAVADKHELLSLASSNHLGKNLQLLMDRVSpécifications
| eukaryotic translation initiation factor 3, subunit H | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003756 | |
| EIF3H | |
| EIF3S3 (NP_003747, 152 a.a. ∼ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 8667 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3B12 | |
| Mouse monoclonal antibody raised against a partial recombinant EIF3S3. | |
| EIF3H | |
| EIF3S3/MGC102958/eIF3-gamma/eIF3-p40 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |