missing translation for 'onlineSavingsMsg'
Learn More

CDK1/CDC2 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Numéro de catalogue. 06966MI
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
06966MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 06966MI Fournisseur MilliporeSigma Code fournisseur 06966
Il en reste null

Rabbit Polyclonal Antibody

Detect Cdk1/Cdc2 with Anti-Cdk1/Cdc2 Antibody, CT (Rabbit Polyclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.

Spécifications

Antigen CDK1/CDC2 (C-Term)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation rabbit antiserum containing 0.05% sodium azide
Gene Symbols CDC2; CDK1; MGC111195; DKFZp686L20222; CDC28A
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a region near the near the C-terminus of human CDK1/CDC2.
Purification Method Antiserum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001777
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Afficher plus de résultats Afficher moins de résultats

For Research Use Only