Similar to CD3 antibody, CD28 antibodies also can be used to stimulate the proliferation and activation of T cells in CIK cell therapy. Under the cooperation of other cytokines, such as IL2 and IL1a, CIK cells with rapid proliferation, high tumoricidal activity, broad tumor killing spectrum and non-MHC-restricted tumor killing characteristics are generatedwhich has significant effects on the treatment of cancer, chronic leukemia, liver disease and neurological diseases. Obviously, It is necessary to control the residues of raw materials in the final cell therapy products.