missing translation for 'onlineSavingsMsg'
Learn More

CD11c Armenian Hamster anti-Mouse, FITC, Clone: N418, MilliporeSigma™

Numéro de catalogue. MABF530MI
Modifier l'affichage
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MABF530MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MABF530MI Fournisseur MilliporeSigma Code fournisseur MABF530
Il en reste null

Armenian Hamster Monoclonal Antibody

This armenian hamster monoclonal antibody Anti-CD11c (mouse), FITC, clone N418 validated for use in immunoprecipitation, immunohistochemistry, Immunofluorescence and Flow Cytometry for the detection of CD11c.

Spécifications

Antigen CD11c
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone N418
Concentration 0.5mg/mL
Conjugate FITC
Formulation Purified Armenian Hamster Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols Itgax
Host Species Armenian Hamster
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 25 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG
Afficher plus de résultats Afficher moins de résultats

For Research Use Only