missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chromosome 1 open reading frame 57 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Description
Sequence: LALPVLRNADCSSGPGQRVCVIDEIGKMELFSQLFIQAVRQTLSTPGTIILGTIPVPKGKPLALVEEIRNRKDVKVFNVTKENRNHLLPDIVTCVQSSRK
Spécifications
Spécifications
| Antigen | chromosome 1 open reading frame 57 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant C1orf57. |
| Formulation | 50% glycerol |
| Gene | C1orf57 |
| Gene Accession No. | NM_032324 |
| Gene Alias | FLJ11383/MGC13186/RP4-678E16.2 |
| Gene Symbols | C1orf57 |
| Afficher plus de résultats |