Learn More
BSA (Bovine Serum Albumin) Rabbit anti-Bovine, Horse, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma 07248
Numéro de catalogue. 07248MI
Il en reste null
Description
Description
This Anti-Albumin Antibody is an antibody against BSA (Bovine Serum Albumin) for use in western blotting.
Spécifications
Spécifications
BSA (Bovine Serum Albumin)
Polyclonal
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Fraction V bovine serum albumin
200 μg
NP_851335.1
−20°C for 24 months
IgG
Western Blot
Unconjugated
Rabbit
Protein A chromatography
Primary
Bovine, Equine, Human, Mouse, Rat
Purified
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS