Learn More
Alpha B-crystallin Rabbit anti-Bovine, Human, Rat, Mouse, Ox, Horse, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma ABN185
Numéro de catalogue. ABN185MI
Il en reste null
Description
Description
This Anti-alpha B-crystallin Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of alpha B-crystallin.
Spécifications
Spécifications
alpha B-crystallin
Polyclonal
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Rabbit
Affinity Purified
Primary
Bovine, Equine, Human, Mouse, Ox, Rat, Rhesus Monkey
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CRYAB; CRYA2
KLH-conjugated linear peptide corresponding to the N-terminus human alpha B-crystallin.
100 μg
NP_001876
2-8°C for 12 months
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS