Learn More
Eisco™ Anthracite Coal Specimen
Catalog No. S24776
Change view
Click to view available options
Quantity:
1 Ea.
Specimen Size:
2 to 3 cm
Catalog No. Quantity Specimen Size
S24776 1 Ea. 2 to 3 cm
1 options
Catalog No. S24776 Supplier Eisco™ Supplier No. ESNG0053
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Anthracite Coal is formed in anoxic environments from compressed organic matter, and is used worldwide as fuel source.
  • Denser than bituminous coal, with higher proportion of carbon
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Closer look at material that powers world
  • Bright and reflective, indicative of intense heat and pressure required to form it
Specifications
Product Type Metamorphic Rock
For Use With (Application) Classroom identification labs and exercises
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Anthracite Coal