missing translation for 'onlineSavingsMsg'
Learn More

Anthracene 97.0+%, TCI America™

Catalog No. p-7116934
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A049525G 25 g
A0495100G 100 g
A0495500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A049525G Supplier TCI America Supplier No. A049525G
Only null left

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 218°C
Boiling Point 342°C
Quantity 25 g
UN Number 3077
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Anthracene
EINECSNumber (4)-0683
RTECSNumber CA9350000
TSCA Yes