missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 99%

Numéro de catalogue. p-60630
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC104860050 5 g Glass Bottle
AC104861000 100 g Plastic bottle
AC104865000 500 g Plastic bottle
AC104860025 2.5 kg Plastic bottle
AC104860100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. AC104860050 Fournisseur Thermo Scientific Chemicals Code fournisseur 104860050
Il en reste null

CAS: 120-12-7 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Spécifications

Melting Point 215.0°C to 218.0°C
Color Yellow
Boiling Point 340.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Beilstein 05,657
Fieser 12,290; 15,99; 16,13
Merck Index 15,674
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/67mL alcohol - 1g/70mL methanol, 1g/200mL ether - 1g/62mL benzene, 1g/85mL chloroform - 1g/31mL carbon disulfide, 1g/86mL ccl4 - 1g/125mL toluene
Formula Weight 178.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Anthracene, 99%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 204-371-1
RTECSNumber CA9350000
TSCA TSCA

RUO – Research Use Only