missing translation for 'onlineSavingsMsg'
Learn More

Anthra[1,9-cd]pyrazol-6(2H)-one 95.0+%, TCI America™

Catalog No. A254825MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A254825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A254825MG Supplier TCI America Supplier No. A254825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129-56-6
Molecular Formula C14H8N2O
Molecular Weight (g/mol) 220.231
MDL Number MFCD00022289
InChI Key ACPOUJIDANTYHO-UHFFFAOYSA-N
Synonym 1,9-pyrazoloanthrone, pyrazolanthrone, dibenzo cd,g indazol-6 2h-one, pyrazoleanthrone, anthra 1,9-cd pyrazol-6 2h-one, anthra-1,9-pyrazol-6-none, 2h-dibenzo cd,g indazol-6-one, jnk inhibitor ii, sapk inhibitor ii
PubChem CID 8515
SMILES C1=CC=C2C(=C1)C3=NNC4=CC=CC(=C43)C2=O

Specifications

Melting Point 282°C
Quantity 25 mg
Formula Weight 220.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Anthra[1,9-cd]pyrazol-6(2H)-one
EINECSNumber (5)-0271
RTECSNumber CB4585000
TSCA Yes
Recommended Storage Refrigerator