Learn More
Ansul™ SPILL-X-FP Formaldehyde Treatment Kit
Free-flowing dry SPILL-X-FP agent to treat spills
Supplier:  Ansul™ 78435
Catalog No. 17-200-6
Only null left
Alert:
The amount of spilled material neutralized or solidified depends on type of acid, caustic, solvent, or formaldehyde and its percentage concentration. Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste materials should follow local, state and federal regulations.
Description
Description
Free-flowing dry SPILL-X-FP agent to treat spills
SPILL-X-FP agent ihas been developed to reach with all formaldehyde solutions. The reaction of SPILL-X-FP agent with aqueous formaldehyde is both temperature and concentration dependent with respect to formaldehyde solutions strenght. Agent components have been formulate to regulate the reaction at an acidic pH, yielding a positive effect on reaction rate.
Order Info
Six single-agent shaker bottles per pack.
Specifications
Specifications
0.80 g/mL density, 2.1 pH
Formaldehyde Spill Control Treatment
Formaldehyde spills
6 Bottles/Pk.
Product Suggestions
Product Suggestions