missing translation for 'onlineSavingsMsg'
Learn More

Ansul™ SPILL-X Multi-Purpose Spill Treatment Kit

Catalog No. 172003
Encompass
Change view
Click to view available options
Packaging Quantity:
6 Bottles/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
172003 6 Bottles/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The amount of spilled material neutralized or solidified depends on type of acid, caustic, solvent, or formaldehyde and its percentage concentration. Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste materials should follow local, state and federal regulations.
Catalog No. 172003 Supplier Ansul™ Supplier No. 77358

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The amount of spilled material neutralized or solidified depends on type of acid, caustic, solvent, or formaldehyde and its percentage concentration. Treated spills containing heavy metals or other listed wastes may still be characterized as hazardous. Disposal of spill waste materials should follow local, state and federal regulations.

Free-flowing dry SPILL-X agent to treat spills

The SPILL-X Multipurpose Spill Treatment Kit contains six (6) handy polypropylene containers with two (2) if each type of SPILL-X spill control agent (SPILL-X-A Acide Neutralizer/Solidifier, SPILL-X-C Caustic Neutralizer/Solidifier, SPILL-X-S Solvent Absorbent).

Order Info

Six single-agent shaker bottles per pack.

Specifications

Color Red, Black, Tan
Type Multi-purpose Spill Control Treatment Kit
For Use With (Application) Acid, caustic, solvent, or formaldehyde spills
Includes 2 bottle of each for SPILL-X-A agent, SPILL-X-C agent, and SPILL-X-S agent
Packaging Quantity 6 Bottles/Pk.