missing translation for 'onlineSavingsMsg'
Learn More

Ansul™ Sentry™ 2 1/2 Water Extinguisher

Catalog No. 189991881
Encompass
Change view
Click to view available options
Discharge Time:
50 sec.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Discharge Time
189991881 50 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189991881 Supplier Ansul™ Supplier No. 430847

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Effective against Class A fire hazards except those with Class C potential (because water conducts electricity). Stainless-steel butt weld shell for long life and corrosion resistance.

UL rating: 2-A. Contains 2 1/2 gal. (9.5L) water. Charged weight: 28 lb. (12.7kg). Dimensions: 7D x 9W x 25 1/2 in.H (17.8 x 22.9 x 64.7cm). One-year limited warranty.

Specifications

Product Type Water Extinguisher
Material Steel
Dimensions (L x W x H) 7 x 9 x 25.5 in. (17.78 x 22.9 x 64.7 cm)
Discharge Time 50 sec.
Height (English) 25.5 in.
Height (Metric) 64.7 cm
Includes Wall hanger
Length (English) 7 in.
Length (Metric) 17.78 cm
Model W02-1
Range 35 ft.
UL Rating 2A
Weight (English) 2.5 lb.
Weight (Metric) 1.13 kg
Width (English) 9 in.
Width (Metric) 22.9 cm
Show More Show Less